siRNA Details
Gene Symbol: NPM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002520.6 | NP_002511.1 | 11 | 1125 | ||
NM_199185.3 | NP_954654.1 | 10 | 1038 | |||
XM_011534564.1 | XP_011532866.1 | 10 | 945 | |||
GenBank mRNA | AK290652.1 | - | 11 | 988 | ||
AY347529.1 | - | 12 | 1015 | |||
AY740634.1 | - | 11 | 956 | |||
AY740635.1 | - | 11 | 956 | |||
AY740636.1 | - | 11 | 956 | |||
AY740637.1 | - | 11 | 956 | |||
AY740640.1 | - | 11 | 949 | |||
BC002398.2 | - | 11 | 967 | 2821577 | ||
BC003670.1 | - | 1 | 682 | |||
BC008495.1 | - | 11 | 977 | 4276604 | ||
BC009623.2 | - | 10 | 919 | 3877633 | ||
BC012566.1 | - | 11 | 980 | 4097025 | ||
BC014349.1 | - | 11 | 976 | 3511037 | ||
BC016716.1 | - | 11 | 979 | 4068167 | ||
BC016768.1 | - | 11 | 979 | 4081364 | ||
BC016824.1 | - | 11 | 979 | 4106286 | ||
BC020467.1 | - | 11 | 966 | |||
BC021668.1 | - | 11 | 980 | 3930893 | ||
BC021983.1 | - | 11 | 978 | 4106023 | ||
BC050628.1 | - | 11 | 966 | 5575414 | ||
BU661768.1 | - | 7 | 614 | |||
DQ303464.1 | - | 11 | 980 | |||
J02590.1 | - | 2 | 244 | |||
M23613.1 | - | 11 | 973 | |||
M26697.1 | - | 11 | 970 | |||
M28699.1 | - | 11 | 967 | |||
M31004.1 | - | 4 | 244 | |||
X16934.1 | - | 11 | 840 |
Gene Symbol: CLEC2D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004419.4 | NP_001004419.1 | 6 | 2037 | ||
NM_001197317.2 | NP_001184246.1 | 4 | 1844 | |||
NM_001197318.2 | NP_001184247.1 | 4 | 1851 | |||
NM_001197319.2 | NP_001184248.1 | 3 | 1740 | |||
NM_013269.5 | NP_037401.1 | 5 | 1955 | |||
NR_036693.2 | 4 | 1926 | ||||
GenBank mRNA | BC094859.1 | - | 1 | 832 |
Gene Symbol: FANCB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017029356.1 | XP_016884845.1 | 11 | 4223 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
