siRNA Details
Gene Symbol: ABCG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AJ300465.1 | - | 7 | 854 | ||
AJ308237.1 | - | 7 | 866 | |||
AJ308244.1 | - | 1 | 53 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
