siRNA Details
Gene Symbol: EPHA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278300.1 | NP_001265229.1 | 13 | 1497 | ||
NM_173655.3 | NP_775926.1 | 13 | 1421 | |||
GenBank mRNA | AL133666.1 | - | 12 | 1368 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
