siRNA Details
Gene Symbol: EPHA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011540969.1 | XP_011539271.1 | 19 | 4616 | ||
XM_011540970.1 | XP_011539272.1 | 17 | 4350 | |||
XM_011540972.1 | XP_011539274.1 | 18 | 4517 | |||
XM_011540973.1 | XP_011539275.1 | 16 | 4252 | |||
XM_011540975.2 | XP_011539277.1 | 14 | 3428 | |||
GenBank mRNA | AB040892.1 | - | 15 | 3874 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
