siRNA Details
Gene Symbol: ORC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002553.3 | NP_002544.1 | 14 | 1903 | ||
GenBank mRNA | AF047599.1 | - | 14 | 1838 | ||
AF049127.1 | - | 14 | 1834 | |||
AK223361.1 | - | 14 | 1903 | |||
AK302122.1 | - | 15 | 1990 | |||
BC023652.2 | - | 14 | 1841 | 4555074 | ||
U92538.1 | - | 14 | 1848 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
