siRNA Details
Gene Symbol: TPM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000366.5 | NP_000357.3 | 9 | 1036 | ||
XM_005254651.1 | XP_005254708.1 | 8 | 862 | |||
XM_017022538.1 | XP_016878027.1 | 9 | 1479 | |||
GenBank mRNA | BC007433.2 | - | 9 | 986 | 2962925 | |
GQ472198.1 | - | 9 | 986 | |||
M19714.1 | - | 9 | 787 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Matching Taqman Gene Expression Assay:
Gene Ontology Categories:
