siRNA Details
Gene Symbol: TPM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301226.1 | NP_001288155.1 | 5, 6 | 799 | ||
NM_003289.3 | NP_003280.2 | 5, 6 | 799 | |||
XM_017015091.1 | XP_016870580.1 | 5, 6 | 671 | |||
GenBank mRNA | BE294047.1 | - | 4, 5 | 360 | ||
M12126.1 | - | 2, 3 | 103 | |||
X06825.1 | - | 5, 6 | 666 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
