siRNA Details
Gene Symbol: REG3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002580.2 | NP_002571.1 | 6 | 539 | ||
NM_138937.2 | NP_620354.1 | 6 | 516 | |||
NM_138938.2 | NP_620355.1 | 5 | 826 | |||
GenBank mRNA | AK291973.1 | - | 6 | 515 | ||
AY544128.1 | - | 5 | 563 | |||
AY544130.1 | - | 5 | 509 | |||
BC036776.1 | - | 5 | 520 | 5749980 | ||
BQ777112.1 | - | 6 | 539 | |||
D13510.1 | - | 5 | 515 | |||
M84337.1 | - | 5 | 520 | |||
S51768.1 | - | 5 | 520 | |||
X68641.1 | - | 5 | 491 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
