siRNA Details
Gene Symbol: CCNH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199189.1 | NP_001186118.1 | 2 | 581 | ||
NM_001239.3 | NP_001230.1 | 2 | 467 | |||
XM_005248627.3 | XP_005248684.1 | 2 | 305 | |||
XM_005248629.4 | XP_005248686.1 | 2 | 643 | |||
XM_011543706.2 | XP_011542008.1 | 2 | 305 | |||
XM_017010014.1 | XP_016865503.1 | 2 | 607 | |||
GenBank mRNA | AB209342.1 | - | 1 | 59 | ||
AK291720.1 | - | 2 | 320 | |||
BC005280.1 | - | 2 | 323 | 3930062 | ||
BC016705.1 | - | 2 | 318 | 4082151 | ||
BC016823.1 | - | 2 | 314 | 4102297 | ||
BC022351.1 | - | 2 | 302 | 4277337 | ||
BP244381.1 | - | 2 | 458 | |||
BX537673.1 | - | 2 | 582 | |||
CR407658.1 | - | 2 | 228 | |||
DA079651.1 | - | 2 | 484 | |||
U11791.1 | - | 2 | 288 | |||
U12685.1 | - | 2 | 240 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
