siRNA Details
Gene Symbol: PGAM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317079.1 | NP_001304008.1 | 3 | 543 | ||
NM_002629.3 | NP_002620.1 | 3 | 613 | |||
GenBank mRNA | AK223096.1 | - | 3 | 459 | ||
AK292216.1 | - | 3 | 514 | |||
AK293023.1 | - | 3 | 458 | |||
AK296619.1 | - | 3 | 402 | |||
AK297491.1 | - | 4 | 359 | |||
AK312351.1 | - | 3 | 442 | |||
AY007118.1 | - | 1 | 450 | |||
BC000455.1 | - | 1 | 476 | 2821567 | ||
BC010038.1 | - | 1 | 468 | 3604026 | ||
BC011678.2 | - | 1 | 432 | 4299008 | ||
BC053356.1 | - | 1 | 449 | 5787678 | ||
BC062302.1 | - | 1 | 448 | 6584466 | ||
BC066959.1 | - | 3 | 481 | 5262935 | ||
BC073742.1 | - | 1 | 447 | 6297173 | ||
BC157873.1 | - | 3 | 465 | 8862699 | ||
CR541659.1 | - | 3 | 430 | |||
CR541744.1 | - | 3 | 430 | |||
EU794644.1 | - | 3 | 442 | |||
J04173.1 | - | 3 | 461 |
Gene Symbol: PGAM4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029891.2 | NP_001025062.1 | 1 | 430 | ||
GenBank mRNA | BG875904.1 | - | 1 | 282 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
