siRNA Details
Gene Symbol: LSS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145436.1 | NP_001138908.1 | 22 | 2954 | ||
NM_001145437.1 | NP_001138909.1 | 21 | 3119 | |||
NM_002340.5 | NP_002331.3 | 22 | 2987 | |||
XM_011529564.2 | XP_011527866.1 | 22 | 3013 | |||
XM_011529566.2 | XP_011527868.1 | 22 | 2934 | |||
XM_011529567.2 | XP_011527869.1 | 21 | 2917 | |||
GenBank mRNA | AK092334.1 | - | 3 | 2337 | ||
AK098352.1 | - | 2 | 473 | |||
AK128839.1 | - | 2698 | ||||
AK226141.1 | - | 21 | 3032 | |||
BC035638.1 | - | 22 | 2950 | 5547173 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
