siRNA Details
Gene Symbol: SERPINB8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276490.1 | NP_001263419.1 | 6 | 2969 | ||
NM_002640.3 | NP_002631.3 | 7 | 3147 | |||
NM_198833.1 | NP_942130.1 | 7 | 3164 | |||
XM_017025792.1 | XP_016881281.1 | 5 | 2771 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
