siRNA Details
Gene Symbol: CDK5R1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003885.2 | NP_003876.1 | 2 | 1469 | ||
XM_011525438.2 | XP_011523740.1 | 2 | 1456 | |||
XM_017025281.1 | XP_016880770.1 | 2 | 2351 | |||
GenBank mRNA | BC020580.2 | - | 2 | 1420 | 4392555 | |
BC026347.1 | - | 1 | 1318 | 4816946 | ||
BC030792.1 | - | 2 | 1475 | 5263087 | ||
BC032081.1 | - | 1 | 748 | |||
BC035448.1 | - | 1 | 748 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
