siRNA Details
Gene Symbol: CERK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022766.5 | NP_073603.2 | 2 | 326 | ||
XM_017028909.1 | XP_016884398.1 | 2 | 123 | |||
GenBank mRNA | AB051433.1 | - | 1 | 52 | ||
AB079066.1 | - | 2 | 342 | |||
AJ457828.1 | - | 2 | 289 | |||
AK291476.1 | - | 2 | 282 | |||
BC067255.1 | - | 2 | 278 | |||
BC126940.1 | - | 2 | 269 | 40084110 | ||
CR456404.1 | - | 2 | 229 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
