siRNA Details
Gene Symbol: CDKL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003948.3 | NP_003939.1 | 9 | 1605 | ||
XM_005263335.2 | XP_005263392.1 | 9 | 1324 | |||
XM_006714406.2 | XP_006714469.1 | 9 | 1324 | |||
XM_017008809.1 | XP_016864298.1 | 9 | 1324 | |||
XM_017008810.1 | XP_016864299.1 | 9 | 1324 | |||
XM_017008811.1 | XP_016864300.1 | 9 | 1324 | |||
GenBank mRNA | AK294866.1 | - | 9 | 1591 | ||
AK309127.1 | - | 2 | 147 | |||
AK312490.1 | - | 9 | 1596 | |||
BC093646.1 | - | 9 | 1129 | 7939491 | ||
BC093981.1 | - | 9 | 1129 | 7939826 | ||
U35146.1 | - | 9 | 1455 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
