siRNA Details
Gene Symbol: PLGLB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002665.4 | NP_002656.1 | 2 | 205 | ||
GenBank mRNA | AK124365.1 | - | 1 | 1109 | ||
BC005379.1 | - | 2 | 201 | 3934860 | ||
BC022294.1 | - | 2 | 207 | 4716526 | ||
M93143.2 | - | 2 | 205 |
Gene Symbol: PLGLA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003506.3 | 2 | 197 | |||
GenBank mRNA | U67178.1 | - | 1 | 56 |
Gene Symbol: PLGLB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001032392.2 | NP_001027564.1 | 2 | 201 | ||
GenBank mRNA | BC070296.1 | - | 2 | 180 | ||
BC107767.1 | - | 2 | 176 | |||
BC109248.2 | - | 2 | 177 | 40022876 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
