siRNA Details
Gene Symbol: PXDNL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144651.4 | NP_653252.3 | 20 | 4044 | ||
XM_005251168.3 | XP_005251225.1 | 6 | 2191 | |||
XM_006716420.3 | XP_006716483.1 | 18 | 5342 | |||
XM_011517456.2 | XP_011515758.1 | 19 | 5558 | |||
XM_011517457.2 | XP_011515759.1 | 19 | 5558 | |||
XM_011517458.2 | XP_011515760.1 | 18 | 3718 | |||
XM_017013041.1 | XP_016868530.1 | 17 | 3679 | |||
GenBank mRNA | AK058200.1 | - | 4 | 1264 | ||
AY877349.1 | - | 20 | 4044 | |||
EU170240.1 | - | 20 | 3943 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
