siRNA Details
Gene Symbol: GCH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000161.2 | NP_000152.1 | 6 | 1259 | ||
XM_017021218.1 | XP_016876707.1 | 6 | 1691 | |||
GenBank mRNA | BC025415.2 | - | 6 | 1246 | 4129035 | |
S44049.1 | - | 6 | 1072 | |||
U19523.1 | - | 6 | 1245 | |||
U66097.1 | - | 6 | 1251 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
