siRNA Details
Gene Symbol: DIO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000792.6 | NP_000783.2 | 1 | 206 | ||
NM_001039715.2 | NP_001034804.1 | 1 | 206 | |||
NM_001039716.2 | NP_001034805.1 | 1 | 206 | |||
NM_001324316.1 | NP_001311245.1 | 1 | 206 | |||
NR_136692.1 | 1 | 206 | ||||
GenBank mRNA | AK290780.1 | - | 1 | 206 | ||
AY560374.1 | - | 1 | 182 | |||
AY560376.1 | - | 1 | 182 | |||
AY560377.1 | - | 1 | 182 | |||
AY560378.1 | - | 1 | 182 | |||
AY560379.1 | - | 1 | 182 | |||
AY560381.1 | - | 1 | 182 | |||
BC017955.1 | - | 1 | 205 | 4607072 | ||
BC107170.2 | - | 1 | 205 | 40036824 | ||
BC107171.2 | - | 1 | 205 | 40036826 | ||
FJ002244.1 | - | 1 | 204 | |||
FJ002245.1 | - | 1 | 204 | |||
FJ002247.1 | - | 1 | 204 | |||
FJ002248.1 | - | 1 | 204 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
