siRNA Details
Gene Symbol: SRGN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK311873.1 | - | 1, 2 | 129 | ||
BC015516.1 | - | 1, 2 | 127 | 3883161 | ||
BP335278.1 | - | 1, 2 | 173 | |||
CB110512.1 | - | 1, 2 | 173 | |||
J03223.1 | - | 1, 2 | 118 | |||
X17042.1 | - | 1, 2 | 111 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
