siRNA Details
Gene Symbol: KYNU | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199241.1 | NP_001186170.1 | 15 | 1722 | ||
NM_003937.2 | NP_003928.1 | 14 | 1636 | |||
XM_011512102.2 | XP_011510404.1 | 13 | 1445 | |||
GenBank mRNA | AK223426.1 | - | 14 | 1634 | ||
AK290900.1 | - | 14 | 1582 | |||
U57721.1 | - | 14 | 1612 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
