siRNA Details
Gene Symbol: RAPGEF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012294.3 | NP_036426.3 | 16 | 1426 | ||
XM_005249914.2 | XP_005249971.1 | 16 | 1645 | |||
XM_005249916.1 | XP_005249973.1 | 16 | 1589 | |||
XM_011515652.2 | XP_011513954.1 | 8 | 3399 | |||
XM_017012837.1 | XP_016868326.1 | 14 | 1330 | |||
GenBank mRNA | AK295859.1 | - | 6 | 911 | ||
BC039203.1 | - | 6 | 737 | 4811483 | ||
BP349215.1 | - | 7 | 484 | |||
D87467.1 | - | 6 | 718 | |||
KU178490.1 | - | 6 | 663 | |||
KU178491.1 | - | 6 | 663 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
