siRNA Details
Gene Symbol: FNIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008738.2 | NP_001008738.2 | 2 | 269 | ||
NM_133372.2 | NP_588613.2 | 2 | 269 | |||
GenBank mRNA | AK292473.1 | - | 2 | 223 | ||
BC001956.1 | - | 2 | 222 | |||
CR936723.1 | - | 2 | 157 | |||
DQ145719.1 | - | 2 | 174 |

More Information
Other Species Targeted

Gene Ontology Categories:
