siRNA Details
Gene Symbol: PSG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | M94891.1 | - | 2 | 302 | ||
U18468.1 | - | 2 | 221 | |||
U18469.1 | - | 2 | 243 | |||
X17097.1 | - | 2 | 308 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
