siRNA Details
Gene Symbol: PSG6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002782.4 | NP_002773.1 | 6 | 1397 | ||
GenBank mRNA | M33666.1 | - | 6 | 1317 |
Gene Symbol: PSG9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017027007.1 | XP_016882496.1 | 6 | 1362 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
