siRNA Details
Gene Symbol: PSG11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113410.1 | NP_001106881.1 | 4, 5 | 785 | ||
NM_002785.2 | NP_002776.3 | 5, 6 | 1151 | |||
GenBank mRNA | BE767872.1 | - | 2, 3 | 254 | ||
BG571293.1 | - | 3, 4 | 493 | |||
M69245.1 | - | 5, 6 | 1113 | |||
U25988.1 | - | 4, 5 | 781 |
Gene Symbol: PSG10P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026824.1 | 6, 7 | 1352 |
Gene Symbol: PSG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031246.3 | NP_112536.2 | 5, 6 | 1216 | ||
GenBank mRNA | BC022316.1 | - | 5, 6 | 1143 | 4731250 | |
M20882.1 | - | 5, 6 | 1127 | |||
M37398.1 | - | 2, 3 | 317 |
Gene Symbol: PSG5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130014.1 | NP_001123486.1 | 5, 6 | 1175 | ||
GenBank mRNA | M25384.1 | - | 6, 7 | 1143 | ||
M73713.1 | - | 5, 6 | 1129 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
