siRNA Details
Gene Symbol: GKAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135953.1 | NP_001129425.1 | 3 | 604 | ||
NM_025211.3 | NP_079487.2 | 3 | 604 | |||
XM_005252241.2 | XP_005252298.1 | 3 | 567 | |||
XM_011519058.2 | XP_011517360.1 | 4 | 462 | |||
GenBank mRNA | AB033131.1 | - | 3 | 400 | ||
AB033132.1 | - | 3 | 400 | |||
AF319476.1 | - | 2 | 282 | |||
AK302100.1 | - | 5 | 633 | |||
BC014476.1 | - | 3 | 509 | 4862778 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
