siRNA Details
Gene Symbol: SOD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000636.3 | NP_000627.2 | 5 | 843 | ||
NM_001322814.1 | NP_001309743.1 | 4 | 726 | |||
NM_001322815.1 | NP_001309744.1 | 4 | 663 | |||
NM_001322819.1 | NP_001309748.1 | 5 | 749 | |||
NM_001322820.1 | NP_001309749.1 | 5 | 752 | |||
GenBank mRNA | AK296809.1 | - | 4 | 639 | ||
AK304766.1 | - | 4 | 576 | |||
M36693.1 | - | 5 | 776 | |||
X07834.1 | - | 5 | 686 | |||
X14322.1 | - | 5 | 777 | |||
X15132.1 | - | 5 | 742 | |||
X59445.1 | - | 5 | 742 | |||
Y00985.1 | - | 5 | 776 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
