siRNA Details
Gene Symbol: SOD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000636.3 | NP_000627.2 | 5 | 918 | ||
NM_001322814.1 | NP_001309743.1 | 4 | 801 | |||
NM_001322815.1 | NP_001309744.1 | 4 | 738 | |||
NM_001322819.1 | NP_001309748.1 | 5 | 824 | |||
NM_001322820.1 | NP_001309749.1 | 5 | 827 | |||
GenBank mRNA | AK296809.1 | - | 4 | 714 | ||
AK304766.1 | - | 4 | 651 | |||
M36693.1 | - | 5 | 851 | |||
X07834.1 | - | 5 | 761 | |||
X14322.1 | - | 5 | 852 | |||
X15132.1 | - | 5 | 817 | |||
X59445.1 | - | 5 | 817 | |||
Y00985.1 | - | 5 | 851 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
