siRNA Details
Gene Symbol: NTMT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286796.1 | NP_001273725.1 | 3 | 686 | ||
NM_001286797.1 | NP_001273726.1 | 3 | 499 | |||
NM_001286798.1 | NP_001273727.1 | 3 | 560 | |||
NM_001286799.1 | NP_001273728.1 | 3 | 489 | |||
NM_001286802.1 | NP_001273731.1 | 2 | 344 | |||
NM_001286803.1 | NP_001273732.1 | 2 | 318 | |||
NM_014064.3 | NP_054783.2 | 3 | 534 | |||
XM_005251939.3 | XP_005251996.1 | 3 | 475 | |||
XM_017014642.1 | XP_016870131.1 | 3 | 392 | |||
GenBank mRNA | AF110776.1 | - | 3 | 477 | ||
AK290957.1 | - | 3 | 531 | |||
AK292332.1 | - | 3 | 536 | |||
AK313829.1 | - | 3 | 524 | |||
BC001396.2 | - | 3 | 513 | 3050940 | ||
BC033234.1 | - | 3 | 488 | 5426447 | ||
BI083998.1 | - | 3 | 499 | |||
BM009113.1 | - | 2 | 271 | |||
BM554138.1 | - | 3 | 489 | |||
BQ923165.1 | - | 3 | 513 | |||
CN406947.1 | - | 3 | 547 | |||
HY058177.1 | - | 2 | 340 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
