siRNA Details
Gene Symbol: PGAM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317079.1 | NP_001304008.1 | 3 | 592 | ||
NM_002629.3 | NP_002620.1 | 3 | 662 | |||
GenBank mRNA | AK223096.1 | - | 3 | 508 | ||
AK292216.1 | - | 3 | 563 | |||
AK293023.1 | - | 3 | 507 | |||
AK295991.1 | - | 4 | 398 | |||
AK296619.1 | - | 3 | 451 | |||
AK297491.1 | - | 4 | 408 | |||
AK312351.1 | - | 3 | 491 | |||
AK315910.1 | - | 3 | 595 | |||
AY007118.1 | - | 1 | 499 | |||
BC000455.1 | - | 1 | 525 | 2821567 | ||
BC010038.1 | - | 1 | 517 | 3604026 | ||
BC011678.2 | - | 1 | 481 | 4299008 | ||
BC053356.1 | - | 1 | 498 | 5787678 | ||
BC062302.1 | - | 1 | 497 | 6584466 | ||
BC066959.1 | - | 3 | 530 | 5262935 | ||
BC073742.1 | - | 1 | 496 | 6297173 | ||
BC157873.1 | - | 3 | 514 | 8862699 | ||
CR541659.1 | - | 3 | 479 | |||
CR541744.1 | - | 3 | 479 | |||
EU794644.1 | - | 3 | 491 | |||
J04173.1 | - | 3 | 510 |
Gene Symbol: PGAM4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029891.2 | NP_001025062.1 | 1 | 479 | ||
GenBank mRNA | BG875904.1 | - | 1 | 331 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
