siRNA Details
Gene Symbol: GUCA1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002098.5 | NP_002089.4 | 4 | 636 | ||
GenBank mRNA | BQ636025.1 | - | 4 | 613 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
