siRNA Details
Gene Symbol: AMPD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001257360.1 | NP_001244289.1 | 3 | 924 | ||
NM_001257361.1 | NP_001244290.1 | 2 | 207 | |||
NM_004037.7 | NP_004028.3 | 2 | 791 | |||
NM_139156.3 | NP_631895.1 | 2 | 571 | |||
GenBank mRNA | AK025706.1 | - | 2 | 547 | ||
AK296394.1 | - | 2 | 163 | |||
AK308907.1 | - | 3 | 397 | |||
AK314494.1 | - | 3 | 847 | |||
BC007711.2 | - | 2 | 496 | 4101667 | ||
BC075844.1 | - | 2 | 496 | 4130690 | ||
DC410161.1 | - | 2 | 207 | |||
U16267.1 | - | 2 | 492 | |||
U16268.1 | - | 2 | 718 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
