siRNA Details
Gene Symbol: ACOT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006821.5 | NP_006812.3 | 3 | 1141 | ||
NR_046028.1 | 4 | 945 | ||||
XM_006720007.1 | XP_006720070.1 | 3 | 628 | |||
GenBank mRNA | AK001939.1 | - | 3 | 1141 | ||
AK093131.1 | - | 4 | 945 | |||
AK223635.1 | - | 3 | 1137 | |||
AY005822.1 | - | 3 | 857 | |||
BC004436.1 | - | 3 | 915 | |||
BC006335.2 | - | 3 | 1125 | 4303771 | ||
BC006500.2 | - | 3 | 915 | |||
DQ082755.1 | - | 3 | 1141 | |||
L40401.1 | - | 3 | 444 |
Gene Symbol: ACOT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037161.1 | NP_001032238.1 | 3 | 971 | ||
XM_017021589.1 | XP_016877078.1 | 3 | 437 | |||
XM_017021590.1 | XP_016877079.1 | 5 | 455 | |||
XM_017021591.1 | XP_016877080.1 | 6 | 938 | |||
XM_017021592.1 | XP_016877081.1 | 3 | 468 | |||
GenBank mRNA | AK300886.1 | - | 3 | 1008 | ||
BC127747.1 | - | 3 | 853 | 40127752 | ||
BC127748.1 | - | 3 | 853 | 40127753 | ||
BC132889.1 | - | 3 | 801 | 40146911 | ||
BC132891.1 | - | 3 | 801 | 40146913 | ||
BC143042.1 | - | 3 | 852 | 40127754 | ||
BC144425.1 | - | 3 | 723 | 9052946 | ||
BX161396.1 | - | 3 | 1081 | |||
DQ082754.1 | - | 3 | 971 | |||
GU727640.1 | - | 3 | 773 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
