siRNA Details
Gene Symbol: ACOT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037161.1 | NP_001032238.1 | 3 | 1281 | ||
XM_017021589.1 | XP_016877078.1 | 3 | 747 | |||
XM_017021590.1 | XP_016877079.1 | 5 | 765 | |||
XM_017021591.1 | XP_016877080.1 | 6 | 1248 | |||
XM_017021592.1 | XP_016877081.1 | 3 | 778 | |||
GenBank mRNA | AK300886.1 | - | 3 | 1318 | ||
BC127747.1 | - | 3 | 1163 | 40127752 | ||
BC127748.1 | - | 3 | 1163 | 40127753 | ||
BC132889.1 | - | 3 | 1111 | 40146911 | ||
BC132891.1 | - | 3 | 1111 | 40146913 | ||
BC143042.1 | - | 3 | 1162 | 40127754 | ||
BC144425.1 | - | 3 | 1033 | 9052946 | ||
BX161396.1 | - | 3 | 1391 | |||
DQ082754.1 | - | 3 | 1281 | |||
GU727640.1 | - | 3 | 1083 |
Gene Symbol: ACOT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006821.5 | NP_006812.3 | 3 | 1451 | ||
NR_046028.1 | 4 | 1255 | ||||
XM_006720007.1 | XP_006720070.1 | 3 | 938 | |||
GenBank mRNA | AK001939.1 | - | 3 | 1451 | ||
AK093131.1 | - | 4 | 1255 | |||
AK223635.1 | - | 3 | 1447 | |||
AY005822.1 | - | 3 | 1167 | |||
BC004436.1 | - | 3 | 1225 | |||
BC006335.2 | - | 3 | 1435 | 4303771 | ||
BC006500.2 | - | 3 | 1225 | |||
DQ082755.1 | - | 3 | 1451 | |||
L40401.1 | - | 3 | 754 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
