siRNA Details
Gene Symbol: HSP90AB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271969.1 | NP_001258898.1 | 3 | 442 | ||
NM_001271970.1 | NP_001258899.1 | 3 | 370 | |||
NM_001271971.1 | NP_001258900.1 | 3 | 429 | |||
NM_001271972.1 | NP_001258901.1 | 3 | 429 | |||
NM_007355.3 | NP_031381.2 | 3 | 429 | |||
GenBank mRNA | AK294643.1 | - | 3 | 286 | ||
AK297367.1 | - | 3 | 286 | |||
AK312255.1 | - | 3 | 290 | |||
AL136543.1 | - | 3 | 265 | |||
AY359878.1 | - | 3 | 303 | |||
BC004928.1 | - | 3 | 275 | 3621040 | ||
BC007327.2 | - | 3 | 267 | |||
BC012807.2 | - | 3 | 275 | 3530042 | ||
BC014485.1 | - | 3 | 270 | 4870198 | ||
BC016753.2 | - | 3 | 288 | 2987963 | ||
BC068474.1 | - | 3 | 293 | 4824686 | ||
BG482327.1 | - | 3 | 264 | |||
BI252490.1 | - | 1 | 276 | |||
BX420114.2 | - | 3 | 330 | |||
BX649079.1 | - | 3 | 282 | |||
CX761569.1 | - | 3 | 319 | |||
DC340766.1 | - | 3 | 362 | |||
DC341882.1 | - | 3 | 378 | |||
DC350708.1 | - | 3 | 429 | |||
DC365600.1 | - | 3 | 364 | |||
M16660.1 | - | 3 | 275 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Matching Taqman Gene Expression Assay:
Gene Ontology Categories:
