siRNA Details
Gene Symbol: HSP90AB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271969.1 | NP_001258898.1 | 6 | 1041 | ||
NM_001271970.1 | NP_001258899.1 | 6 | 969 | |||
NM_001271971.1 | NP_001258900.1 | 6 | 884 | |||
NM_001271972.1 | NP_001258901.1 | 6 | 998 | |||
NM_007355.3 | NP_031381.2 | 6 | 1028 | |||
GenBank mRNA | AF275719.1 | - | 4 | 514 | ||
AK025370.1 | - | 3 | 331 | |||
AK290734.1 | - | 1 | 147 | |||
AK294643.1 | - | 6 | 855 | |||
AK297367.1 | - | 6 | 771 | |||
AK312255.1 | - | 6 | 889 | |||
AL136543.1 | - | 6 | 864 | |||
AY359878.1 | - | 6 | 902 | |||
BC004928.1 | - | 6 | 874 | 3621040 | ||
BC007327.2 | - | 6 | 866 | |||
BC009206.2 | - | 4 | 568 | |||
BC012807.2 | - | 6 | 874 | 3530042 | ||
BC014485.1 | - | 6 | 869 | 4870198 | ||
BC016753.2 | - | 6 | 887 | 2987963 | ||
BC068474.1 | - | 6 | 892 | 4824686 | ||
BX649079.1 | - | 6 | 891 | |||
M16660.1 | - | 6 | 874 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
