siRNA Details
Gene Symbol: PLGLB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002665.4 | NP_002656.1 | 4 | 809 | ||
GenBank mRNA | AK124365.1 | - | 3 | 1713 | ||
BC005379.1 | - | 4 | 805 | 3934860 | ||
M93143.2 | - | 4 | 809 |
Gene Symbol: PLG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168338.1 | NP_001161810.1 | 4 | 855 |
Gene Symbol: PLGLA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003506.3 | 4 | 802 | |||
GenBank mRNA | U67178.1 | - | 3 | 661 |
Gene Symbol: PLGLB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001032392.2 | NP_001027564.1 | 4 | 805 | ||
GenBank mRNA | BC070296.1 | - | 4 | 784 | ||
BC107767.1 | - | 4 | 780 | |||
BC109248.2 | - | 4 | 781 | 40022876 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
