siRNA Details
Gene Symbol: ADCY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281768.1 | NP_001268697.1 | 4 | 578 | ||
NM_021116.2 | NP_066939.1 | 3 | 892 | |||
XM_005249584.3 | XP_005249641.1 | 3 | 890 | |||
XM_005249585.2 | XP_005249642.1 | 3 | 890 | |||
GenBank mRNA | AB209496.1 | - | 3 | 837 | ||
AF497515.1 | - | 3 | 896 | |||
L05500.1 | - | 1 | 36 |
