siRNA Details
Gene Symbol: SLC9A3R2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130012.2 | NP_001123484.1 | 6, 7 | 992 | ||
NM_001252073.1 | NP_001239002.1 | 5, 6 | 609 | |||
NM_001252075.1 | NP_001239004.1 | 5, 6 | 603 | |||
XM_006720981.1 | XP_006721044.1 | 5, 6 | 2469 | |||
XM_017023895.1 | XP_016879384.1 | 6, 7 | 847 | |||
GenBank mRNA | AF004900.1 | - | 6, 7 | 870 | ||
AF035771.1 | - | 6, 7 | 854 | |||
BC014513.1 | - | 6, 7 | 965 | |||
BC069014.1 | - | 6, 7 | 961 | |||
BC106001.1 | - | 6, 7 | 968 | 5296143 | ||
BX377861.2 | - | 5, 6 | 595 | |||
CB961565.1 | - | 5, 6 | 630 | |||
Z50150.1 | - | 6, 7 | 947 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
