siRNA Details
Gene Symbol: PMS2P7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_130940.1 | 3, 4 | 324 | |||
GenBank mRNA | AB017006.1 | - | 3, 4 | 419 | ||
DB472687.1 | - | 3, 4 | 324 |
Gene Symbol: PMS2P3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028059.1 | 6, 7 | 1085 |
Gene Symbol: PMS2P5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | DW407867.1 | - | 3, 4 | 325 |
Gene Symbol: PMS2P9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BM845848.1 | - | 2, 3 | 127 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
