siRNA Details
Gene Symbol: SLC35A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042498.2 | NP_001035963.1 | 4 | 1287 | ||
NM_001282649.1 | NP_001269578.1 | 3 | 1104 | |||
NM_001282650.1 | NP_001269579.1 | 4 | 1326 | |||
NM_001282651.1 | NP_001269580.1 | 5 | 1371 | |||
NM_005660.2 | NP_005651.1 | 4 | 1287 | |||
GenBank mRNA | AK090730.1 | - | 3 | 536 | ||
AK225508.1 | - | 4 | 975 | |||
AK292816.1 | - | 4 | 1004 | |||
AK293415.1 | - | 3 | 792 | |||
AK293419.1 | - | 4 | 1014 | |||
AK298484.1 | - | 5 | 1059 | |||
CR450287.1 | - | 4 | 965 | |||
D84454.1 | - | 4 | 1288 | |||
D88146.1 | - | 4 | 969 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
