siRNA Details
Gene Symbol: SLCO1B3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019844.3 | NP_062818.1 | 4 | 346 | ||
GenBank mRNA | AB669023.1 | - | 2 | 132 | ||
AF187815.1 | - | 2 | 105 | |||
AJ251506.1 | - | 2 | 149 | |||
AK055874.1 | - | 4 | 324 | |||
AY342017.1 | - | 2 | 119 | |||
BC022843.1 | - | 3 | 230 | |||
BC058006.1 | - | 3 | 235 | |||
BC070264.1 | - | 3 | 209 | |||
BC105597.1 | - | 3 | 235 | |||
BG567966.1 | - | 3 | 232 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
