siRNA Details
Gene Symbol: SLC39A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144564.4 | NP_653165.2 | 3 | 592 | ||
GenBank mRNA | AF052125.1 | - | 1 | 116 | ||
AK302600.1 | - | 3 | 533 | |||
BC005869.2 | - | 3 | 524 | 2966557 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
