siRNA Details
Gene Symbol: SLC22A23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286456.1 | NP_001273385.1 | 1, 2 | 650 | ||
NM_015482.1 | NP_056297.1 | 1, 2 | 650 | |||
GenBank mRNA | AK308943.1 | - | 1, 2 | 134 | ||
BC040876.1 | - | 1, 2 | 592 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
