siRNA Details
Gene Symbol: SLC22A23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286455.1 | NP_001273384.1 | 9 | 1148 | ||
NM_015482.1 | NP_056297.1 | 9 | 1607 | |||
NM_021945.6 | NP_068764.3 | 10 | 1393 | |||
NR_104448.1 | 10 | 1201 | ||||
XM_011514801.2 | XP_011513103.1 | 10 | 1694 | |||
XM_017011180.1 | XP_016866669.1 | 10 | 2171 | |||
XM_017011181.1 | XP_016866670.1 | 8 | 920 | |||
XM_017011182.1 | XP_016866671.1 | 7 | 1202 | |||
XM_017011184.1 | XP_016866673.1 | 7 | 857 | |||
XM_017011186.1 | XP_016866675.1 | 10 | 1090 | |||
GenBank mRNA | AK172770.1 | - | 6 | 703 | ||
AK308282.1 | - | 10 | 1146 | |||
AK308943.1 | - | 9 | 1091 | |||
BC038748.1 | - | 10 | 1364 | |||
BC128580.1 | - | 9 | 1100 | 40128787 | ||
BC128581.1 | - | 10 | 1157 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
