siRNA Details
Gene Symbol: SLC22A23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286455.1 | NP_001273384.1 | 3, 4 | 448 | ||
NM_001286456.1 | NP_001273385.1 | 3, 4 | 907 | |||
NM_015482.1 | NP_056297.1 | 3, 4 | 907 | |||
NM_021945.6 | NP_068764.3 | 4, 5 | 693 | |||
NR_104448.1 | 3, 4 | 448 | ||||
XM_011514801.2 | XP_011513103.1 | 4, 5 | 994 | |||
XM_017011180.1 | XP_016866669.1 | 4, 5 | 1471 | |||
XM_017011181.1 | XP_016866670.1 | 2, 3 | 220 | |||
XM_017011186.1 | XP_016866675.1 | 4, 5 | 390 | |||
GenBank mRNA | AK308282.1 | - | 4, 5 | 447 | ||
AK308943.1 | - | 3, 4 | 391 | |||
AL110245.1 | - | 2, 3 | 351 | |||
BC038748.1 | - | 4, 5 | 664 | |||
BC040876.1 | - | 3, 4 | 849 | |||
BC128580.1 | - | 3, 4 | 400 | 40128787 | ||
BC128581.1 | - | 3, 4 | 404 | |||
DB050701.1 | - | 3, 4 | 449 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
