siRNA Details
Gene Symbol: PSG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276495.1 | NP_001263424.1 | 5 | 1314 | ||
NM_001316339.1 | NP_001303268.1 | 5 | 1227 | |||
NM_002780.4 | NP_002771.2 | 6 | 1593 | |||
NM_213633.2 | NP_998798.1 | 5 | 1314 | |||
XM_017026996.1 | XP_016882485.1 | 7 | 1655 | |||
XM_017027000.1 | XP_016882489.1 | 6 | 1380 | |||
GenBank mRNA | AF289606.1 | - | 6 | 934 | ||
AK075149.1 | - | 6 | 1208 | |||
BC008405.1 | - | 8 | 1452 | 4043484 | ||
BC063127.1 | - | 6 | 1461 | 4768860 | ||
M33665.1 | - | 4 | 734 | |||
M94891.1 | - | 6 | 1441 | |||
U18468.1 | - | 6 | 1360 | |||
U18469.1 | - | 5 | 1103 | |||
X17097.1 | - | 6 | 1447 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
