siRNA Details
Gene Symbol: PSG7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290042.1 | NP_001276971.1 | 2 | 452 | ||
NM_002783.2 | NP_002774.2 | 2 | 452 | |||
GenBank mRNA | AL546656.3 | - | 1 | 29 | ||
BC030979.1 | - | 2 | 447 | |||
BG434892.1 | - | 2 | 449 | |||
BP321474.1 | - | 2 | 451 | |||
BX097470.1 | - | 1 | 74 | |||
BX952498.1 | - | 2 | 450 | |||
U18467.1 | - | 2 | 402 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
