siRNA Details
Gene Symbol: PGAM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317079.1 | NP_001304008.1 | 4 | 1104 | ||
NM_002629.3 | NP_002620.1 | 4 | 1174 | |||
GenBank mRNA | AK223096.1 | - | 4 | 1020 | ||
AK292216.1 | - | 4 | 1075 | |||
AK293023.1 | - | 4 | 1019 | |||
AK295991.1 | - | 5 | 910 | |||
AK297491.1 | - | 5 | 920 | |||
AK315910.1 | - | 4 | 1107 | |||
AY007118.1 | - | 1 | 1011 | |||
BC000455.1 | - | 1 | 1037 | 2821567 | ||
BC010038.1 | - | 1 | 1029 | 3604026 | ||
BC011678.2 | - | 1 | 993 | 4299008 | ||
BC053356.1 | - | 1 | 1010 | 5787678 | ||
BC062302.1 | - | 1 | 1009 | 6584466 | ||
BC066959.1 | - | 4 | 1042 | 5262935 | ||
BC073742.1 | - | 1 | 1008 | 6297173 | ||
BC157873.1 | - | 4 | 1026 | 8862699 | ||
J04173.1 | - | 4 | 1022 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
